Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Et-29 | 2166487-23-4 | 99.9% | C34H46N6O6S | 10MG
SDP

Supplier:  Medchemexpress LLC HY14565110MG

Encompass_Preferred

Et-29 is a potent and selective inhibitor of sirtuin 5 (SIRT5) with a reported Ki of 40 nM. Provided for research use, the compound is characterized by high purity, defined solubility in DMSO, and recommended cold storage to preserve stability.

  • Potent and selective SIRT5 inhibition (Ki = 40 nM).
  • High reported purity (99.89%).
  • Soluble in DMSO at high concentration (100 mg/mL); may require ultrasonic.
  • Solid, white to off-white appearance.
  • Recommended storage under nitrogen at -20°C; in solution, long-term storage at -80°C.
  • Available in multiple small milligram pack sizes for research use.

Catalog No. 50-002-58383


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.