Learn More
Lonza Walkersville Endothelin-3 Growth Supplement (ET-3)
Supplier:  Lonza Walkersville CC4510
Encompass
ET-3 Growth Supplement is an additional growth factor required for adult melanocyte growth. It is sold separately to supplement the MGM-4 BulletKit (catalog number CC-3249) for a complete formulation specifically designed for growing adult melanocytes. ET-3 offers enhanced growth and plating advantages in addition to limiting other cell type contamination in culture.Content:1 x Lyophilized Vial Containing 130 ug
Catalog No. NC0874902