Learn More
TARGETMOL CHEMICALS INC ETAMICASTAT HYDROCHLORID 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11238L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Etamicastat hydrochloride (BIA 5-453 hydrochloride) is a peripherally selective dopamine beta-hydroxylase inhibitor that reduces hypertension. purity: 99%
Catalog No. 50-258-1545