Learn More
Medchemexpress LLC Etamicastat hydrochloride | 677773-32-9 | 98.14% | 347.81 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14838A10MM/1ML
Encompass_Preferred
Etamicastat hydrochloride (BIA 5-453 hydrochloride) is a potent and reversible dopamine-β-hydroxylase (DBH) inhibitor with an IC50 value of 107 nM. It can be utilized in research pertaining to cardiovascular diseases.
  • Potent and reversible dopamine-β-hydroxylase (DBH) inhibitor
  • IC50 value of 107 nM
  • Useful in cardiovascular disease research
  • Supplied as 10 mM solution in DMSO
  • Storage: Solid at 4°C under nitrogen; In solvent, -80°C for 6 months or -20°C for 1 month under nitrogen.
Catalog No. 50-003-60173