Learn More
Selleck Chemical LLC Etanercept P1240-5MG
SDP
Supplier:  Selleck Chemical LLC P12405MG
Encompass_Preferred
Etanercept a dimeric fusion protein functions as an inhibitor of TNF by binding to TNF It competitively blocks the attachment of both TNF- and TNF- to their cell surface receptors thereby neutralizing TNF s biological activity Etanercept has proven effectiveness in treating rheumatoid arthritis juvenile idiopathic arthritis and plaque psoriasis
Catalog No. 50-000-39761