Learn More
Medchemexpress LLC HY-108847 1mg , Etanercept CAS:185243-69-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1088471MG
Encompass_Preferred
HY-108847 1mg Etanercept CAS: 185243-69-0 Etanercept, a dimeric fusion protein that binds TNF, acts as a TNF inhibitor. Etanercept competitively inhibits the binding of both TNF-α and TNF-β to cell surface TNF receptors, rendering TNF biologically inactive. Etanercept shows efficacy against rheumatoid arthritis, juvenile idiopathic arthritis, and plaque psoriasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9625