Learn More
TARGETMOL CHEMICALS INC ETANERCEPT 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T374455MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Etanercept is a dimeric fusion protein that serves as a competitive TNF inhibitor by binding to TNF. It effectively inhibits the binding of both TNF-(alpha) and TNF-(beta) to the TNF receptors on cell surfaces thereby rendering TNF biologically inactive. Etanercept has demonstrated efficacy in treating rheumatoid arthritis juvenile idiopathic arthritis and plaque psoriasis. purity: 98%
Catalog No. 50-258-0138