Learn More
Selleck Chemical LLC ETC-1002 5mg 738606-46-7 Bempedoic acid, ESP-55016
SDP
Supplier:  Selleck Chemical LLC S79535MG
Encompass_Preferred
ETC-1002 (Bempedoic acid, ESP-55016),also known as Bempedoic acid, is an orally available, once-daily LDL-C lowering small molecule designed to lower elevated levels of LDL-C and to avoid side effects associated with existing LDL-C lowering therapies.ETC-1002 is an activator of hepatic AMP-activated protein kinase (AMPK). It has potent inhibitory activity against hepatic ATP-citrate lyase(IC50=29 uM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5471