Learn More
Alpco Ltd ETEC/EIEC PCR Kit
SDP
Supplier:  Alpco Ltd 86ETIHU100
Encompass
The ETEC/EIEC PCR Kit is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of the virulence-factor genes of ETEC and EIEC/Shigella spp. in human stool and culture supernatant samples. This kit is sufficient for 100 tests.
Catalog No. NC2284604