Learn More
Medchemexpress LLC HY-P1955A 5mg Medchemexpress, Etelcalcetide (hydrochloride) CAS:1334237-71-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1955A5MG
Encompass_Preferred
Medchemexpress, HY-P1955A 5mg Etelcalcetide (HCl) CAS:1334237-71-6 Etelcalcetide hydrochloride (AMG 416 hydrochloride) is a synthetic peptide as an activator of the calcium sensing receptor (CaSR). Etelcalcetide hydrochloride is effective in lowering parathyroid hormone (PTH) concentrations in patients receiving dialysis with secondary hyperparathyroidism receiving hemodialysis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0548