Learn More
Medchemexpress LLC Etelcalcetide (hydrochloride) | 1334237-71-6 | 99.8% | 1353.33 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1955A10MG
Encompass_Preferred
Etelcalcetide hydrochloride is a synthetic calcimimetic that acts as an activator of the calcium sensing receptor (CaSR). It is effective in lowering parathyroid hormone (PTH) concentrations in patients receiving dialysis with secondary hyperparathyroidism, showing promise for research in secondary hyperparathyroidism and chronic kidney disease.
  • Synthetic calcimimetic
  • Activator of the calcium sensing receptor (CaSR)
  • Lowers parathyroid hormone (PTH) concentrations
  • Supports research in secondary hyperparathyroidism
  • Supports research in chronic kidney disease
Catalog No. 50-004-22576