Learn More
Medchemexpress LLC Eteplirsen | 1173755-55-9 | 98.5% | 10306.00 | 1 ML
Supplier: Medchemexpress LLC HY1087531MG
Eteplirsen (AVI 4658) is a synthetic antisense oligonucleotide. It is used for Duchenne muscular dystrophy research. It promotes dystrophin production by restoring the translational reading frame of Duchenne muscular dystrophy (DMD) through specific skipping of exon 51 in defective gene variants.
- Synthetic antisense oligonucleotide
- Can be used for Duchenne muscular dystrophy research
- Promotes dystrophin production by restoring the translational reading frame through specific skipping of exon 51