Learn More
Selleck Chemical LLC Ethacrynate Sodium
SDP
Supplier:  Selleck Chemical LLC S58475MG
Encompass_Preferred
Ethacrynate Sodium (ethacrynic acid sodium) is the sodium salt form of ethacrynic acid which inhibits symport of sodium potassium and chloride primarily in the ascending limb of Henle but also in the proximal and distal tubules
Catalog No. 50-000-03833