Learn More
Cayman Chemical EthacrynIc AcId 1g
SDP
Supplier:  Cayman Chemical 195361 G
Encompass_Preferred
A loop diuretic with anticancer activity; inhibits the NKCC cotransporter in duck erythrocytes (IC50 = 0.18 mM); inhibits ATP-dependent chloride uptake in rat renal plasma membrane vesicles at 0.3 mM; inhibits GSTP1-1 and GSTA3-3 (IC50s = 4.9 and ~0.4 μM, respectively); inhibits Wnt/β-catenin signaling in a cell-based reporter assay; cytotoxic to primary chronic lymphocytic leukemia cells (IC50 = 8.56 μM), as well as MCF-7, MDA-MB-231, and 4T1 cancer cells (IC50s = 45.53, 39.64, and 25.23 μM, respectively); increases tumor growth reduction induced by afatinib or neratinib in a 4T1 murine breast cancer model at 250 μg per day
Catalog No. 50-298-4394