Learn More
Cayman Chemical EthacrynIc AcId 50mg
SDP
Supplier:  Cayman Chemical 2629950 MG
Encompass_Preferred
An analytical reference standard categorized as a loop diuretic; has been used as a masking agent in sports doping, including racehorse doping; intended for use in analytical forensic applications
Catalog No. 50-293-5080