Learn More
Medchemexpress LLC Ethacrynic acid | 58-54-8 | 98.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1640R5MG
Encompass_Preferred
Ethacrynic acid (Standard) is an analytical standard of Ethacrynic acid, used in research and analytical applications. It functions as a diuretic and an inhibitor of glutathione S-transferases (GSTs). It also potently inhibits the NF-kB-signaling pathway, modulates leukotriene formation, and inhibits L-type voltage-dependent and store-operated calcium channels, leading to relaxation of airway smooth muscle (ASM) cells.
  • Functions as a diuretic
  • Inhibits glutathione S-transferases (GSTs)
  • Potently inhibits the NF-kB-signaling pathway
  • Modulates leukotriene formation
  • Inhibits L-type voltage-dependent and store-operated calcium channels
  • Reduces retinoid-induced ear edema in mice (anti-inflammatory)
  • Used as a reference standard for assays
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-13295