Learn More
Medchemexpress LLC Ethacrynic acid | 58-54-8 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB164050MG
Encompass_Preferred
Ethacrynic acid is an orally active diuretic with anti-inflammatory and anticancer activity. It functions as an inhibitor of glutathione S-transferase (GSTs) and Wnt signaling pathways. It can also act as a radiosensitizer, inhibit airway smooth muscle contraction, and increase aqueous humor outflow from the eye.
  • Exhibits anti-inflammatory and anticancer activity.
  • Inhibits Wnt/β-catenin signaling.
  • Cytotoxic in CLL cells.
  • Increases aqueous humor outflow facility.
  • Inhibits activation of NF-κB pathway.
  • Increases radiation intensity in cancer cells.
  • Inhibits tracheal ring contraction.
  • Decreases intracellular Ca2+ concentration.
  • Inhibits tumor growth in mice models.
Catalog No. 50-004-12134