Learn More
Medchemexpress LLC Ethacrynic acid | 58-54-8 | 99.98% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1640100MG
Encompass_Preferred
Ethacrynic acid is an orally active diuretic with anti-inflammatory and anticancer activity. It is an inhibitor of glutathione S-transferase (GSTs) and Wnt signaling pathways, and also functions as a radiosensitizer. Ethacrynic acid can inhibit airway smooth muscle (ASM) contraction in mice and can increase the outflow of aqueous humor from the eye for the study of glaucoma.
  • Anti-inflammatory activity
  • Anticancer activity
  • Orally active diuretic
  • Inhibitor of glutathione S-transferase (GSTs) and Wnt signaling pathways
  • Radiosensitizer
  • Can inhibit airway smooth muscle (ASM) contraction in mice
  • Can increase the outflow of aqueous humor from the eye for the study of glaucoma
Catalog No. 50-004-13198