Learn More
Medchemexpress LLC Ethacrynic acid (Standard) | 58-54-8 | 98.8% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB1640R
Encompass_Preferred
Ethacrynic acid (Standard) is the analytical standard of Ethacrynic acid, intended for research and analytical applications. It is a diuretic and an inhibitor of glutathione S-transferases (GSTs). It also acts as a potent inhibitor of the NF-κB-signaling pathway, modulates leukotriene formation, and inhibits L-type voltage-dependent and store-operated calcium channels, leading to relaxation of airway smooth muscle (ASM) cells. This compound possesses anti-inflammatory properties that reduce retinoid-induced ear edema in mice.
  • Analytical standard
  • Intended for research and analytical applications
  • Diuretic
  • Inhibitor of glutathione S-transferases (GSTs)
  • Potent inhibitor of NF-κB-signaling pathway
  • Modulates leukotriene formation
  • Inhibits L-type voltage-dependent and store-operated calcium channel
  • Anti-inflammatory properties (reduces retinoid-induced ear edema in mice)
Catalog No. 50-004-14052