Learn More
Medchemexpress LLC Ethambutol dihydrochloride | 1070-11-7 | 99.9% | 1 ML
Supplier: Medchemexpress LLC HYB0535A10MM/1ML
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. It is also used to establish animal models of hyperuricemia and optic neuropathy.
- Antimycobacterial compound
- Inhibits arabinosyltransferase activity
- Purity of 99.9%
- Used in hyperuricemia animal models
- Used in optic neuropathy animal models
- Solid, white to off-white appearance