Learn More
Medchemexpress LLC Ethambutol dihydrochloride | 1070-11-7 | 99.9% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0535AR
Encompass_Preferred
Ethambutol dihydrochloride is an antimycobacterial compound that prevents cell wall formation by inhibiting arabinosyltransferase activity. It can be used to establish animal models of hyperuricemia and optic neuropathy.
  • Antimycobacterial compound
  • Prevents cell wall formation
  • Inhibits arabinosyltransferase activity
  • Establishes animal models of hyperuricemia
  • Establishes animal models of optic neuropathy
Catalog No. 50-004-07056