Learn More
Medchemexpress LLC Ethanamine, 2-(2-azidoethoxy)- | 464190-91-8 | ≥98.0% | C4H10N4O | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140212
Encompass_Preferred
Azido-PEG1-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Reacts via copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts via strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-52860