Learn More
Medchemexpress LLC Ethanamine, 2-(phenylmethoxy)- | 38336-04-8 | MFCD00235180 | 99.7% | 151.21 g/mol | C9H13NO | 100 G

Supplier: Medchemexpress LLC HYW017834100G
2-(Benzyloxy)-1-ethanamine (2-(benzyloxy)ethylamine) is an organic biochemical reagent used as a building block and intermediate in organic synthesis and life-science research. It is supplied with high reported purity and is suitable for derivatization, functionalization, and other synthetic transformations; consult the batch certificate of analysis for exact specifications and handling guidance.
- High purity (≈99.7%) suitable for synthetic and research applications.
- Molecular formula C9H13NO, molecular weight 151.21 g/mol.
- Commonly used as an intermediate or building block in organic synthesis.
- Available in multiple pack sizes, including 100 g, for scale flexibility.
- Refer to batch COA for exact purity, analytical data, and handling instructions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.