Learn More
Medchemexpress LLC Ethanaminium, 2-hydrazinyl-N,N,N-trimethyl-2-oxo-, chloride (1:1) | 123-46-6 | MFCD00012009 | 99.8% | 167.64 g/mol | C5H14ClN3O | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW074541
Encompass_Preferred
Girard's reagent T is a hydrazide-based derivatizing reagent that reacts with aldehydes and ketones to form hydrazones, improving detection sensitivity and chromatographic behavior for analytical methods such as mass spectrometry and HPLC.
  • High purity, 99.8%.
  • Forms stable, water-soluble hydrazones with carbonyl compounds.
  • Suitable for mass spectrometry and chromatographic analysis.
  • Molecular weight 167.64 g/mol and formula C5H14ClN3O.
  • Available as a 25 g crystalline powder pack.
Catalog No. 50-002-09162