Learn More
Medchemexpress LLC Ethanedioic acid, 1,2-bis(2,4,6-trichlorophenyl) ester | 1165-91-9 | MFCD00043061 | ≥98.0% | 448.90 g/mol | C14H4Cl6O4 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW042438
Encompass_Preferred
Bis(2,4,6-trichlorophenyl) oxalate is a white to off-white solid oxalate ester used as a biochemical assay reagent and as an organic compound for life-science research. It has molecular formula C14H4Cl6O4, a molecular weight of 448.90 g/mol, and is supplied at high purity for laboratory applications.
  • High purity (≥98.0%).
  • White to off-white solid appearance.
  • Available in multiple sizes including 25 g.
  • Store solid at room temperature; stable for up to 3 years.
  • Suitable for use as a chemiluminescence reagent and organic synthesis intermediate.
Catalog No. 50-002-09752