Learn More
Medchemexpress LLC Ethanesulfonic-1,2-13C2 acid, 2-(amino-15N)- | 2483830-42-6 | 98.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0351S21MG
Encompass_Preferred
Taurine-13C2,15N is a 13C- and 15N-labeled form of Taurine, a sulfur-containing amino acid and organic osmolyte. It plays a role in cell volume regulation, serves as a substrate for bile salt formation, and modulates intracellular free calcium concentration. Additionally, it can activate autophagy in adipocytes.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07208