Learn More
Medchemexpress LLC Ethanesulfonic acid, 2-[(2-amino-2-oxoethyl)amino]- | 7365-82-4 | 98.0% | 100 G
SDP
Supplier:  Medchemexpress LLC HYD0866100G
Encompass_Preferred
ACES (N-(2-Acetamido)-2-aminoethanesulfonic acid) is a zwitterionic buffer primarily used for research purposes. Its working pH range is 6.8-7.2.
  • Functions as a zwitterionic buffer
  • Offers a working pH range of 6.8-7.2
  • Intended for research use only
  • Not validated for medical applications
Catalog No. 50-004-09962