Learn More
Medchemexpress LLC Ethanol, 2-[2-[2-[2-[(2,4-dinitrophenyl)amino]ethoxy]ethoxy]ethoxy]- | 1807520-99-5 | 97.9% | C14H21N3O8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140618100MG
Encompass_Preferred
DNP-PEG4-alcohol is a chemical compound commonly utilized as a PROTAC linker, playing a crucial role in proteolysis-targeting chimera (PROTAC) technology. This molecule is designed to facilitate targeted protein degradation by connecting a ligand that binds to a target protein with another ligand that recruits an E3 ubiquitin ligase. The PEG4 chain enhances solubility and provides conformational flexibility to optimize PROTAC efficacy.
  • Acts as a PROTAC linker
  • Facilitates targeted protein degradation
  • Enhances solubility
  • Provides conformational flexibility
Catalog No. 50-003-53057