Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethoxy]-, 1-(4-methylbenzenesulfonate) | 236754-49-7 | 98.5% | 417.48 g/mol | C17H27N3O7S | 50 MG
SDP

Supplier:  Medchemexpress LLC HY14035250MG

Encompass_Preferred

Azide-PEG5-Tos is an azide-functionalized polyethylene glycol linker (PEG length = 5 units) with a tosylate leaving group, used for bioconjugation and assembly of antibody-drug conjugates. It enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) reactions to attach payloads to alkyne-bearing molecules.

  • Contains an azide moiety for click chemistry conjugations.
  • Peg5 spacer provides hydrophilicity and flexibility.
  • Tosylate group serves as a reactive leaving group for functionalization.
  • High purity suitable for research applications.
  • Soluble in DMSO at ≥100 mg/mL for convenient stock solutions.

Catalog No. 50-002-04060


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.