Learn More
Medchemexpress LLC Ethanol, 2-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethoxy] tosylate | 236754-49-7 | 99.3% | 417.48 g/mol | C17H27N3O7S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140352500MG
Encompass_Preferred
Azide-PEG5-Tos is a cleavable PEG-based linker bearing an azide functional group and a tosylate leaving group. It is used as a click-chemistry handle and linker in the synthesis of antibody-drug conjugates and other bioconjugation applications. The reagent is supplied as a solid and is typically dissolved in DMSO for reactions.
  • Cleavable 5-unit polyethylene glycol spacer
  • Contains an azide functional group for copper-catalyzed azide-alkyne cycloaddition
  • Contains a tosylate leaving group for downstream modification
  • High purity suitable for synthetic chemistry applications
  • Stable under cold storage when handled per supplier recommendations
Catalog No. 50-002-03783