Learn More
Medchemexpress LLC Ethanol, 2,2'-[iminobis(2,1-ethanediyloxy)]bis- | 54384-47-3 | MFCD00239212 | 97.0% | 193.24 g/mol | C8H19NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130328100MG
Encompass_Preferred
NH-bis-PEG2 is a short, non-cleavable two-unit polyethylene glycol (PEG) linker with terminal amine and hydroxyl functionality, used as a hydrophilic spacer in bioconjugation chemistry such as antibody-drug conjugate and PROTAC linker synthesis.
  • Short, flexible two-unit PEG spacer for bioconjugation.
  • Non-cleavable linker with amine and hydroxyl termini.
  • High purity: 97.0% suitable for synthetic applications.
  • Colorless to light yellow liquid with density ≈ 1.09 g/cm3.
  • Stable when stored cold and protected from light; stable in solvent at -80°C for up to 6 months.
Catalog No. 50-002-02606