Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanol, 2-[(phenylmethyl)amino]- | 104-63-2 | MFCD00002840 | 99.3% | 151.21 g/mol | C9H13NO | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW017395

Encompass_Preferred

2-(Benzylamino)ethanol (N-benzylethanolamine) is an organic intermediate and biochemical reagent intended for research use. It is used as a building block in organic synthesis and as a reagent in biochemical assays, and is supplied with manufacturer guidance on purity, storage, and handling.

  • High purity (99.3%) for reliable experimental results.
  • Chemical formula C9H13NO and molecular weight 151.21 g/mol.
  • CAS number 104-63-2 for unambiguous identification.
  • Typical pack size 25 g for laboratory use.
  • Storage: store at 4°C and protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
  • For research use only; not validated for clinical applications.

Catalog No. 50-003-04207


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.