Learn More
Medchemexpress LLC Ethanol, 2-[(tetrahydro-2H-pyran-2-yl)oxy]- | 2162-31-4 | MFCD00185588 | 99.5% | 146.18 | C7H14O3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03877110G
Encompass_Preferred
THP-PEG1-alcohol is a THP-protected PEG-based linker with a primary alcohol functional group, intended for use in the synthesis of PROTACs and other linker-containing small molecules. The reagent provides a short, flexible spacer and a THP-protected hydroxyl handle that can be selectively deprotected in multistep synthetic sequences, facilitating modular assembly of bifunctional compounds.
  • Short PEG spacer suitable for compact linker design.
  • THP-protected hydroxyl enables selective deprotection strategies.
  • High purity supports reproducible synthesis and analytical results.
  • Low molecular weight for minimal added bulk to conjugates.
  • Available in small lab-scale quantities for rapid prototyping.
Catalog No. 50-002-99804