Learn More
HCL Labels Ethanol 70% GHS Label - 3" x 5" Adhesive Vinyl (Pack of 25)
SDP
Supplier:  HCL Labels GH21750VN0035
Encompass_Preferred
Ethanol 70% 3" x 5" (Pack of 25) GHS Compliant Chemical Safety Label for secondary containers. Made from a dependable 3M adhesive vinyl that is laminated for chemical and solvent resistance. HCL's GHS labels have been tested to withstand acetone, dichloromethane, hydrofluoric acid, and other tough chemicals and solvents. These labels are used for containers that store hazardous substances in order to provide employees with the right-to-know of the immediate chemical hazards. Our GHS labels are compliant with OSHA HazCom standard 1910.1200 and aligned with the Globally Harmonized System for identifying and classifying chemical hazards. HCL's GHS safety labels are made in the United States of America.
Catalog No. 50-111-7508