Learn More
Bioworld Ethanol, Denatured, 81%, 500 mL
Supplier:  Bioworld 405700111
Encompass_Preferred
It is a volatile, flammable, colorless straight-chain, primary alcohol,used as a solvent, and as a fuel. Appearance: Colourless liquidAtomic Number: Not available.Boiling Point : 78°CChEBI : Not available.Crystal_Structure : Not available.Density : 0.789 g/cm3Electron Configuration : Not available.InChI : Not available.InChIKey : Not available.Ionization Energy : Not available.IUPAC_Name: EthanolMelting Point: W22;114°CMolar Mass: Not available.NFPA 704: Not available.RTECS Number: KQ6300000Synonyms: Ethyl Alcohol; Reagent Alcohol
Catalog No. 50-255-966
 Restricted Item