Learn More
SEKISUI DIAGNOSTICS LLC ETHANOL - L3K ASSAY KIT
Supplier:  SEKISUI DIAGNOSTICS LLC 27330
Encompass_Preferred
Reagent for the IN VITRO quantitative measurement of ethanol concentration in serum, plasma, and urine.
Catalog No. NC9764936