Learn More
Alkali Scientific Ethanolamine, 4 Liter
SDP
Supplier:  Alkali Scientific HAN002CP
Encompass_Preferred
Ethanolamine is an organic amine compound commonly used in biochemical and industrial applications for buffer preparation and pH adjustment. It acts as a weak base and is frequently utilized in the formulation of biological buffers and chemical solutions where controlled pH is required. In laboratory settings, ethanolamine is used in protein purification processes, surface coating reactions, and as a component in chromatographic and analytical methods. It also plays a role in lipid metabolism studies and membrane research due to its structural relevance in phospholipids. The liquid form allows easy handling and precise measurement for preparation of reagents in research and laboratory workflows.
Catalog No. 50-006-59674