Learn More
Medchemexpress LLC Ethanolamine (hydrochloride) | 2002-24-6 | MFCD00012892 | ≥99.0% | 97.54 g·mol⁻¹ | C2H8ClNO | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW035903
Encompass_Preferred
Ethanolamine (hydrochloride) is the hydrochloride salt of ethanolamine used as a laboratory reagent for buffer preparation, pH adjustment, and biochemical assays. It is a crystalline, water-soluble salt provided with quality documentation for research use.
  • High purity reagent suitable for buffer preparation and assay formulation.
  • Crystalline solid, easy to weigh and dissolve in water.
  • Molecular formula C2H8ClNO; molecular weight 97.54 g·mol⁻¹.
  • Available in small pack sizes for laboratory use.
  • Supplied with certificate of analysis and safety data sheet.
Catalog No. 50-003-03556