Learn More
Medchemexpress LLC Ethanone, 1-[(1'R)-1'-(4-chloro-2-methoxyphenyl)-5'-fluoro-1',2',3',9'-tetrahydrospiro[cyclopropane-1,4'-[4H]pyrido[3,4-b]indol]-2'-yl]- | 2988224-39-9 | 99.81% | 398.86 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY111191A10MG
Encompass_Preferred
(R)-ONO-2952 is the R-enantiomer of ONO-2952, which is a potent, selective, and orally active TSPO antagonist. It exhibits Ki values between 0.33-9.30 nM for rat and human TSPO. ONO-2952 demonstrates greater selectivity for TSPO compared to other receptors, transporters, ion channels, and enzymes.
  • R-enantiomer of ONO-2952.
  • Potent, selective, and orally active TSPO antagonist.
  • Exhibits Ki values of 0.33-9.30 nM for rat and human TSPO.
  • More selective for TSPO than other receptors, transporters, ion channels, and enzymes.
Catalog No. 50-003-43586