Learn More
Medchemexpress LLC Ethanone, 1-[2-(1-pyrrolidinylmethyl)-1-piperidinyl]-2-[4-(trifluoromethyl)phenyl] hydrochloride | 115730-24-0 | 100.0% | 390.87 | C19H26ClF3N2O | 50 MG

Supplier: Medchemexpress LLC HY10158250MG
ZT 52656A hydrochloride is the hydrochloride salt of a selective kappa opioid receptor agonist described for ocular analgesia. It is supplied as a solid research reagent for laboratory and preclinical studies and is not for human or clinical use.
- Selective kappa opioid receptor agonist suitable for ocular pain studies.
- High reported purity (99.98%) for consistent experimental results.
- Solid, white to off-white form for easy handling and weighing.
- Molecular weight 390.87 and formula C19H26ClF3N2O for reference.
- Recommended storage: 4°C sealed; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
- Packaged quantities suitable for small-scale laboratory experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.