Learn More
Medchemexpress LLC Ethanone, 1-(2,4-dimethylphenyl)- | 89-74-7 | MFCD00003571 | 98.9% | 148.20 g/mol | C10H12O | 100 G
SDP
Supplier:  Medchemexpress LLC HYW015618100G
Encompass_Preferred
2',4'-Dimethylacetophenone (CAS 89-74-7) is a small aromatic ketone used as a research chemical and an endogenous metabolite reference. It is supplied in both solid form and ready-to-use DMSO solution and is used as an analytical standard, synthesis intermediate, and in metabolomics research.
  • High purity (98.88%).
  • Available as solid and 10 mM solution in DMSO, multiple pack sizes.
  • Suitable as an analytical standard and synthesis intermediate.
  • Molecular formula C10H12O, molecular weight 148.20 g/mol.
  • Supplied with quality documentation, including COA and SDS.
Catalog No. 50-003-00070