Learn More
Medchemexpress LLC Ethanone, 1-(2,4-dimethylphenyl)- | 89-74-7 | MFCD00003571 | 98.9% | 148.20 g·mol⁻¹ | C10H12O | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01561850G
Encompass_Preferred
2',4'-Dimethylacetophenone (CAS 89-74-7) is an acetophenone derivative reported as an endogenous metabolite and supplied for research use. It is used as a reference compound in biochemical and analytical assays and as an intermediate in organic synthesis. The compound has well-characterized physical properties and high reported purity suitable for laboratory applications.
  • High purity (98.88%) suitable for analytical applications.
  • Molecular formula C10H12O and molecular weight 148.20 g·mol⁻¹ for accurate calculations.
  • Colorless to light yellow liquid with density 0.997 g/cm³.
  • Supplied in multiple pack sizes for flexible laboratory use.
  • Useful as a reference standard and reaction intermediate in organic synthesis.
Catalog No. 50-003-00050