Learn More
Medchemexpress LLC Ethanone, 1-(2,5-dihydroxyphenyl)-, 2-(tetrahydrobenzothieno[2,3-d]pyrimidin-4-yl)hydrazone | 301655-16-3 | 98.0% | 354.43 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY171579
Encompass_Preferred
RS47 is a selective RelB inhibitor with a Kd value of 1.1 μM. It also acts as an inhibitor of HCV replication and can block the non-canonical NF-κB signaling pathway without affecting the canonical pathway. RS47 has anti-tumor effects, inhibiting proliferation and promoting apoptosis in colorectal cancer, B-cell lymphoma, and other related tumors both in vitro and in vivo by disrupting the binding of RelB to target DNA. It can be used for the research of tumors and infectious diseases.
  • Selective RelB inhibitor (Kd value of 1.1 μM)
  • Inhibits HCV replication
  • Blocks non-canonical NF-κB signaling pathway
  • Exerts anti-tumor effects (inhibiting proliferation, promoting apoptosis)
  • Disrupts binding of RelB to target DNA
  • Suitable for research on tumors and infectious diseases
Catalog No. 50-003-85338