Learn More
Medchemexpress LLC Ethanone, 1-(4-amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1) | 37845-71-9 | 98.3% | 311.64 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY33450100MG
Encompass_Preferred
Clenbuterol impurity 1, with the chemical name Ethanone, 1-(4-amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1), is a high-purity chemical compound. It possesses a molecular weight of 311.64 and a molecular formula of C12H17Cl3N2O. This compound typically appears as a white to off-white solid, making it suitable for a range of laboratory research applications.
  • High purity: 98.26% (HPLC)
  • White to off-white solid appearance
  • Structure verified by ¹H NMR spectrum
  • Stable with recommended storage at room temperature for 3 years, or in solvent at -80°C for 2 years, -20°C for 1 year
Catalog No. 50-004-04050