Learn More
Medchemexpress LLC Ethanone, 1-(4-aminophenyl)- | 99-92-3 | 99.97% | 135.16 | 100 G

Supplier: Medchemexpress LLC HYW008818100G
1-(4-Aminophenyl)ethanone, also known as 4'-Aminoacetophenone, is a biochemical reagent primarily used for life science-related research as a biological material or organic compound. It is provided as a yellow to brown solid.
- Consistent with structure by ¹H NMR spectrum and LCMS.
- Highly pure, with a GC purity of 99.97%.
- Store at 4°C, protected from light.
- Soluble in DMSO, 40% PEG300, 20% SBE-β-CD in saline, and corn oil.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.