Learn More
Medchemexpress LLC EthD (bromide) | 1239-45-8 | 99.3% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYD002110MM/1ML
Encompass_Preferred
EthD bromide is an intercalating agent commonly used as a fluorescent tag (nucleic acid stain) in molecular biology laboratories for techniques such as agarose gel electrophoresis.
  • Fluorescent tag (nucleic acid stain) for molecular biology laboratories.
  • Used in techniques such as agarose gel electrophoresis.
  • Allows visualization of DNA under ultraviolet (UV) light.
Catalog No. 50-004-10567