Learn More
Medchemexpress LLC EthD-III | 95.0% | 978.36 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYD17231MG
Encompass_Preferred
EthD-III is a nucleic acid probe that serves as a red fluorescent stain for detecting dead cells. It functions by entering cells with damaged membranes and binding to nucleic acids, which results in bright red fluorescence (Ex/Em=530/645 nm) in dead cells. This product is intended for research use only.
  • Nucleic acid probe
  • Red fluorescent stain
  • Dead cell detection
  • Fluorescence properties
  • Research use only
Catalog No. 50-004-31242