Learn More
Medchemexpress LLC EthD-III | 95.0% | 978.36 | 100 UG
SDP
Supplier:  Medchemexpress LLC HYD1723100UG
Encompass_Preferred
EthD-III is a nucleic acid probe. It is a red fluorescent stain used to detect dead cells. This probe enters cells with damaged membranes and binds to nucleic acids, producing bright red fluorescence (Ex/Em=530/645 nm).
  • Detects dead cells
  • Enters cells with damaged membranes
  • Binds to nucleic acids
  • Produces bright red fluorescence
Catalog No. 50-004-29994