Learn More
Echelon Biosciences Research Labs ETHER C18 LBPA 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs LB1B1250UG
Encompass
Biotin-C12-ether LBPA is a biotin conjugate of ether C12 Lysobisphosphatidic acid ((R,R)-2,2-Bisdodecyl-LBPA ammonium salt), an ether analog of the naturally occurring, biologically active isomer of LPBA.Lysobisphosphatidic acids (LBPAs), known also as bis-(monoacylglycerol)phosphates (BMPs) are specialized lipid molecules reported to play a role in intracellular protein and lipid transport in healthy cells. Their accumulation was observed in pathologic liver tissue, and they also serve as antigens for auto-antibody generation in a human autoimmune condition termed antiphospholipid syndrome. Accumulation of LBPAs in intracellular, often multilamellar membranes is related to biomembrane polymorphism which may impact intracellular cholesterol transport. This item is non returnable
Catalog No. 50-720-541