Learn More
AdipoGen Ethidium homodimer,Chemical. CAS: 61926-22-5
SDP
Supplier:  AdipoGen CDXE0012M005
Encompass_Preferred
Ethidium homodimer, AdipoGen Life Sciences. Chemical. CAS: 61926-22-5. Formula: C46H50Cl4N8. MW: 856.75. Synthetic. Staining dye for ssDNA, dsDNA, RNA, oligonucleotides and triplex DNA. It does not cross intact cell membranes and can be used to test cell viability. Reagent for the fluorimetric detection of nucleic acids.
Catalog No. 50-101-6645